Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
phospho2
Entrez Gene ID
799638
Gene Name
phosphatase, orphan 2
Gene Aliases
zgc:172082
Chromosome Location
Chr.6: 3710628 - 3713117 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111229.1 NP_001104699.1 1 - 1 1943 120
GenBank mRNA BC155600.1 - 1 - 1 1943 120

View Full Product Details