Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pi4kab
Entrez Gene ID
793349
Gene Name
phosphatidylinositol 4-kinase, catalytic, alpha b
Gene Aliases
zgc:172062
Chromosome Location
Chr.8: 1741932 - 1838315 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113342.1 NP_001106813.1 3 - 4 367 70
GenBank mRNA BC155592.1 - 3 - 4 367 70

View Full Product Details