Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trim32
Entrez Gene ID
562764
Gene Name
tripartite motif containing 32
Gene Aliases
-
Chromosome Location
Chr.Unknown: 29380 - 35204 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113594.1 NP_001107066.1 - 2151 63
XM_005174681.2 XP_005174738.1 - 2255 63
GenBank mRNA BC154641.1 - - 2151 63
BC171687.1 - - 2145 63
BC171691.1 - - 2145 63

View Full Product Details