Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
znfl1b
Entrez Gene ID
795031
Gene Name
zinc finger-like gene 1b
Gene Aliases
si:dkey-103d23.5, znfl1, znfl1d
Chromosome Location
Chr.9: 15837398 - 15841997 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113633.1 NP_001107105.1 1 - 2 377 114

View Full Product Details