Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
il6r
Entrez Gene ID
100135755
Gene Name
interleukin 6 receptor
Gene Aliases
il-6ra
Chromosome Location
Chr.16: 22177254 - 22194400 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330258.2 NP_001317187.2 6 - 7 1762 71

View Full Product Details