Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hoxb8a
Entrez Gene ID
30343
Gene Name
homeobox B8a
Gene Aliases
fj67g02, gp-8, hoxb8, wu:fj67g02, zgc:64159
Chromosome Location
Chr.3: 23687838 - 23690657 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131120.1 NP_571195.1 1 - 2 800 74
GenBank mRNA BC053287.1 - 1 - 2 800 74
DQ060543.1 - 1 - 2 256 74

View Full Product Details