Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
oprm1
Entrez Gene ID
65088
Gene Name
opioid receptor, mu 1
Gene Aliases
ZFOR2, or2
Chromosome Location
Chr.13: 23843614 - 23882977 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131707.2 NP_571782.2 2 - 3 729 101
GenBank mRNA AF132813.1 - 2 - 3 638 101

View Full Product Details