Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
efnb2b
Entrez Gene ID
114402
Gene Name
ephrin-B2b
Gene Aliases
fc10d03, hm:zeh0344, wu:fc10d03, zeh0344
Chromosome Location
Chr.1: 7634516 - 7659569 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131808.1 NP_571883.1 2 - 3 407 101
GenBank mRNA AF375226.1 - 2 - 3 407 101

View Full Product Details