Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbx6
Entrez Gene ID
192294
Gene Name
T-box transcription factor 6
Gene Aliases
ZF-TBX24, id:ibd5070, tbx24, tbx6r, z.TBX24
Chromosome Location
Chr.12: 3912544 - 3923070 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153666.2 NP_705952.2 5 - 6 878 86
GenBank mRNA AB036516.1 - 5 - 6 878 86
BC163297.1 - 5 - 6 766 86

View Full Product Details