Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
adssl
Entrez Gene ID
192305
Gene Name
adenylosuccinate synthase, like
Gene Aliases
adss, adss2, chunp6912, zgc:66318
Chromosome Location
Chr.21: 26522638 - 26531032 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173237.2 NP_775344.2 7 - 8 824 65
GenBank mRNA AY099534.1 - 7 - 8 698 65
BC051613.1 - 7 - 8 686 65

View Full Product Details