Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glula
Entrez Gene ID
100000775
Gene Name
glutamate-ammonia ligase (glutamine synthase) a
Gene Aliases
cb195, cb757, etID49849.2, fb39g09, fd19d09, gso1, sb:cb195, wu:fb39g09, wu:fd19d09
Chromosome Location
Chr.2: 18988410 - 18992347 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181559.2 NP_853537.1 6 - 7 915 91
GenBank mRNA BC045255.1 - 6 - 7 913 91
BC053146.1 - 6 - 7 900 91
BC171365.1 - 5 - 6 831 91
BC171369.1 - 6 - 7 836 91

View Full Product Details