Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glulb
Entrez Gene ID
336473
Gene Name
glutamate-ammonia ligase (glutamine synthase) b
Gene Aliases
fb11d08, gso2, hm:zehn1949, im:7153900, wu:fb11d08
Chromosome Location
Chr.6: 32195 - 37427 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182866.3 NP_878286.3 4 - 5 924 94
GenBank mRNA BC045886.1 - 4 - 5 596 94
BC066735.1 - 4 - 5 581 94
BC164204.1 - 3 - 4 509 94
BC168499.1 - 4 - 5 596 94

View Full Product Details