Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp4t8
Entrez Gene ID
387527
Gene Name
cytochrome P450, family 4, subfamily T, polypeptide 8
Gene Aliases
cb1041, cyp4
Chromosome Location
Chr.16: 237325 - 280350 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC056734.1 - 10 - 11 1282 142

View Full Product Details