Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
asb8
Entrez Gene ID
327200
Gene Name
ankyrin repeat and SOCS box containing 8
Gene Aliases
wu:fd16d08, zgc:64033
Chromosome Location
Chr.23: 27497734 - 27506180 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199770.1 NP_956064.1 3 - 4 320 73
XM_005162226.4 XP_005162283.1 3 - 4 590 73
XM_005162227.4 XP_005162284.1 3 - 4 378 73
GenBank mRNA BC053213.1 - 3 - 4 320 73
BC153632.1 - 3 - 4 347 73
BC165704.1 - 2 - 3 266 73

View Full Product Details