Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
polr2c
Entrez Gene ID
334669
Gene Name
RNA polymerase II subunit C
Gene Aliases
wu:fk48d01, zgc:56285
Chromosome Location
Chr.25: 14369496 - 14378869 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167956.2 NP_001161428.1 3 - 4 395 70
GenBank mRNA BC046047.1 - 3 - 4 307 70
EH555113.1 - 3 - 4 439 70

View Full Product Details