Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
txndc9
Entrez Gene ID
336625
Gene Name
thioredoxin domain containing 9
Gene Aliases
fb15e07, wu:fb15e07, zgc:55522
Chromosome Location
Chr.9: 188132 - 193637 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328575.1 NP_001315504.1 2 - 3 278 85
NM_001328576.1 NP_001315505.1 2 - 3 235 85
NM_200021.2 NP_956315.1 2 - 3 281 85
GenBank mRNA BC045369.1 - 2 - 3 282 85
BC067598.1 - 1 - 2 212 85
BC165424.1 - 1 - 2 225 85
EH491524.1 - 2 - 3 250 85

View Full Product Details