Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ctu1
Entrez Gene ID
393098
Gene Name
cytosolic thiouridylase subunit 1 homolog (S. pombe)
Gene Aliases
zgc:55395
Chromosome Location
Chr.8: 3423148 - 3439490 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200130.1 NP_956424.1 2 - 3 732 65
XM_005172440.4 XP_005172497.3 2 - 3 793 65
GenBank mRNA BC044158.1 - 2 - 3 732 65
BC154793.1 - 2 - 3 730 65
BC164148.1 - 1 - 2 532 65

View Full Product Details