Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nol12
Entrez Gene ID
393260
Gene Name
nucleolar protein 12
Gene Aliases
zgc:56457
Chromosome Location
Chr.3: 1150376 - 1156033 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200290.1 NP_956584.1 2 - 3 258 68
GenBank mRNA BC049449.1 - 2 - 3 258 68

View Full Product Details