Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aprt
Entrez Gene ID
393641
Gene Name
adenine phosphoribosyltransferase
Gene Aliases
zgc:65965
Chromosome Location
Chr.7: 55530533 - 55539728 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200668.1 NP_956962.1 2 - 3 202 72
GenBank mRNA BC058046.1 - 2 - 3 202 72
BC155778.1 - 2 - 3 212 72

View Full Product Details