Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bcl6aa
Entrez Gene ID
393707
Gene Name
BCL6A transcription repressor a
Gene Aliases
bcl6, bcl6a, zgc:73162
Chromosome Location
Chr.6: 173459 - 179500 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200734.1 NP_957028.1 6 - 7 1751 64
XM_017356456.2 XP_017211945.1 6 - 7 1783 64
XM_021476813.1 XP_021332488.1 7 - 8 1871 64
XM_021476814.1 XP_021332489.1 6 - 7 1737 64
XM_021476815.1 XP_021332490.1 6 - 7 2046 64
GenBank mRNA BC059515.1 - 6 - 7 1751 64

View Full Product Details