Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arid3c
Entrez Gene ID
569444
Gene Name
AT rich interactive domain 3C (BRIGHT-like)
Gene Aliases
arid3a, si:ch211-113n10.6
Chromosome Location
Chr.5: 380576 - 382734 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100039.1 NP_001093509.1 7 - 8 1536 76
XM_005165358.4 XP_005165415.1 8 - 9 1998 76

View Full Product Details