Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyth4b
Entrez Gene ID
100141338
Gene Name
cytohesin 4b
Gene Aliases
zgc:175224
Chromosome Location
Chr.6: 392817 - 409768 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114903.2 NP_001108375.2 2 - 3 128 111
GenBank mRNA BC154839.1 - 2 - 3 108 111
EH453546.1 - 2 - 3 159 111

View Full Product Details