Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sept8a
Entrez Gene ID
321590
Gene Name
septin 8a
Gene Aliases
fb22a03, sept8, wu:fb22a03
Chromosome Location
Chr.21: 43328732 - 43374893 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115117.1 NP_001108589.1 3 - 4 411 122
GenBank mRNA BC055257.1 - 3 - 4 440 122
BC081662.1 - 3 - 4 411 122
BC100044.1 - 3 - 4 406 122
BC151876.1 - 3 - 4 425 122

View Full Product Details