Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bnc2
Entrez Gene ID
559327
Gene Name
basonuclin 2
Gene Aliases
bnp, coral, crl, fi18b05, poppy, si:dkey-114f6.1, wu:fi18b05
Chromosome Location
Chr.1: 27023908 - 27209699 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001327750.1 NP_001314679.1 4 - 5 824 109
NM_001327751.1 NP_001314680.1 4 - 5 941 109
NM_001327752.1 NP_001314681.1 4 - 5 910 109
XM_017354977.2 XP_017210466.1 4 - 5 1101 109
GenBank mRNA BC157380.1 - 4 - 5 882 109
GQ229411.1 - 3 - 4 440 109

View Full Product Details