Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc6a22.2
Entrez Gene ID
561024
Gene Name
solute carrier family 6 member 22, tandem duplicate 2
Gene Aliases
SLC6A6, si:ch211-263a23.6, si:zc263a23.6, slc6a13l
Chromosome Location
Chr.6: 41144142 - 41161021 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122761.1 NP_001116233.1 10 - 11 1368 123
XM_005166045.4 XP_005166102.1 10 - 11 1381 123
XM_005166046.4 XP_005166103.1 11 - 12 1290 123

View Full Product Details