Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhp2
Entrez Gene ID
100003787
Gene Name
NHP2 ribonucleoprotein homolog (yeast)
Gene Aliases
nola2, wu:fb36b10, wu:fd61a03, zgc:73227
Chromosome Location
Chr.21: 30401780 - 30408763 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212597.2 NP_997762.1 1 - 2 283 82
GenBank mRNA BC051784.1 - 1 - 2 234 82
BC059569.1 - 1 - 2 229 82
EH485076.1 - 1 - 2 278 82
EH537394.1 - 1 - 2 305 82
EH553640.1 - 1 - 2 317 82
EH557479.1 - 3 - 4 668 82

View Full Product Details