Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smndc1
Entrez Gene ID
572679
Gene Name
survival motor neuron domain containing 1
Gene Aliases
TDRD10, spf30, wu:fb37h07, wu:fc23a07
Chromosome Location
Chr.17: 220515 - 230237 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212601.2 NP_997766.1 3 - 4 440 72
GenBank mRNA BC045967.1 - 3 - 4 423 72
BC067338.1 - 3 - 4 383 72
BC165076.1 - 2 - 3 298 72
EH547286.1 - 3 - 4 448 72
EH552076.1 - 3 - 4 438 72
EH588730.1 - 3 - 4 427 72

View Full Product Details