Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbcb
Entrez Gene ID
337350
Gene Name
tubulin folding cofactor B
Gene Aliases
ckap1, ik:tdsubc_2e4, wu:fa56d03, xx:tdsubc_2e4, zgc:55620
Chromosome Location
Chr.18: 8405 - 19345 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212775.1 NP_997940.1 5 - 6 777 90
GenBank mRNA BC044438.1 - 5 - 6 777 90
BC153663.1 - 5 - 6 773 90
BC164828.1 - 4 - 5 588 90

View Full Product Details