Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
esrra
Entrez Gene ID
405891
Gene Name
estrogen-related receptor alpha
Gene Aliases
err1, erra, erralpha, esrral
Chromosome Location
Chr.21: 26606672 - 26636691 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212955.1 NP_998120.1 2 - 3 505 75
XM_005161344.4 XP_005161401.1 3 - 4 757 75
XM_009295473.3 XP_009293748.1 4 - 5 1061 75
GenBank mRNA AY556395.1 - 2 - 3 505 75

View Full Product Details