Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
esrra
Entrez Gene ID
405891
Gene Name
estrogen-related receptor alpha
Gene Aliases
err1, erra, erralpha, esrral
Chromosome Location
Chr.21: 26606672 - 26636691 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212955.1 NP_998120.1 5 - 6 1051 72
XM_005161344.4 XP_005161401.1 6 - 7 1303 72
XM_009295473.3 XP_009293748.1 7 - 8 1607 72
GenBank mRNA AY556395.1 - 5 - 6 1051 72

View Full Product Details