Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsf1a
Entrez Gene ID
406288
Gene Name
serine and arginine rich splicing factor 1a
Gene Aliases
sfrs1a, sfrs1l, wu:fb52g10, wu:fb97g12, zgc:66146
Chromosome Location
Chr.15: 80347 - 84615 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213015.1 NP_998180.1 3 - 4 456 93
XM_005157414.4 XP_005157471.1 3 - 4 544 93
XM_005157415.4 XP_005157472.1 3 - 4 592 93
GenBank mRNA BC055511.1 - 3 - 4 488 93
BC153596.1 - 3 - 4 482 93

View Full Product Details