Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppib
Entrez Gene ID
406292
Gene Name
peptidylprolyl isomerase B (cyclophilin B)
Gene Aliases
cb87, sb:cb87, wu:fa97f08, zgc:73214, zgc:86796
Chromosome Location
Chr.25: 5983448 - 5990182 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213019.1 NP_998184.1 3 - 4 500 79
GenBank mRNA BC059560.1 - 3 - 4 367 79
BC071458.1 - 3 - 4 500 79
BC154213.1 - 3 - 4 391 79

View Full Product Details