Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hnrnpl2
Entrez Gene ID
573096
Gene Name
heterogeneous nuclear ribonucleoprotein L2
Gene Aliases
fb53f10, fp61c04, hnrnpl, hnrpl, wu:fb53f10, wu:fp61c04, zgc:66175
Chromosome Location
Chr.15: 87549 - 102335 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213383.1 NP_998548.1 1 - 2 128 89
XM_005169692.4 XP_005169749.1 1 - 2 149 89
XM_005169693.4 XP_005169750.1 1 - 2 149 89
XM_017350996.2 XP_017206485.1 1 - 2 149 89
GenBank mRNA BC054655.1 - 1 - 2 128 89
BC153481.1 - 1 - 2 100 89
BC154328.1 - 1 - 2 91 89
BC165353.1 - 1 - 2 116 89

View Full Product Details