Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hnrnpl2
Entrez Gene ID
573096
Gene Name
heterogeneous nuclear ribonucleoprotein L2
Gene Aliases
fb53f10, fp61c04, hnrnpl, hnrpl, wu:fb53f10, wu:fp61c04, zgc:66175
Chromosome Location
Chr.15: 87549 - 102335 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213383.1 NP_998548.1 2 - 3 253 69
XM_005169692.4 XP_005169749.1 2 - 3 274 69
XM_005169693.4 XP_005169750.1 2 - 3 274 69
XM_017350996.2 XP_017206485.1 2 - 3 274 69
GenBank mRNA BC054655.1 - 2 - 3 253 69
BC154328.1 - 2 - 3 216 69
BC165353.1 - 2 - 3 241 69

View Full Product Details