Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bsx
Entrez Gene ID
573364
Gene Name
brain-specific homeobox
Gene Aliases
-
Chromosome Location
Chr.10: 99786 - 101618 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214727.1 NP_999892.1 1 - 2 338 91
GenBank mRNA AY515251.1 - 1 - 2 338 91
BC162183.1 - 1 - 2 313 91
BC162209.1 - 1 - 2 313 91

View Full Product Details