Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- capn2l
- Entrez Gene ID
-
445091
- Gene Name
- calpain 2, (m/II) large subunit, like
- Gene Aliases
-
CH73-80N6.1, wu:fb09d04, wu:fb26a06 gene, xx:hac23b03, zgc:112420, zgc:92480
- Chromosome Location
-
Chr.22: 26443235 - 26476079 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC093295.1 |
- |
4 - 5 |
587 |
127 |
BC154357.1 |
- |
4 - 5 |
573 |
127 |
View Full Product Details