Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
si:dkey-286h2.3
Entrez Gene ID
790965
Gene Name
si:dkey-286h2.3
Gene Aliases
cyp2x12
Chromosome Location
Chr.7: 156772 - 165382 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079853.2 NP_001073322.1 9 - 9 1644 136
GenBank mRNA AY825257.1 - 9 - 9 1645 136
BC117654.1 - 9 - 9 1628 136

View Full Product Details