Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL1R2
Entrez Gene ID
100033831
Gene Name
interleukin 1 receptor type 2
Gene Aliases
IL-1RII
Chromosome Location
Chr.15: 7832890 - 7868572 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081816.1 NP_001075285.1 1 - 2 66 72
GenBank mRNA AB033415.1 - 1 - 2 66 72

View Full Product Details