Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL1RN
Entrez Gene ID
100034236
Gene Name
interleukin 1 receptor antagonist
Gene Aliases
IL-1RA
Chromosome Location
Chr.15: 16702120 - 16716417 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082525.2 NP_001075994.1 2 - 3 262 146
XM_005599766.3 XP_005599823.1 - 220 146
XM_014730883.2 XP_014586369.1 - 303 146
GenBank mRNA D83714.1 - 2 - 3 216 146
U92482.1 - 2 - 3 216 146

View Full Product Details