Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA12
Entrez Gene ID
100054028
Gene Name
carbonic anhydrase 12
Gene Aliases
-
Chromosome Location
Chr.1: 130007880 - 130057701 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099760.1 NP_001093230.1 5 - 6 539 77
XM_005602598.3 XP_005602655.2 - 836 77
GenBank mRNA EF397505.1 - 5 - 6 539 77

View Full Product Details