Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELENOP
Entrez Gene ID
100052968
Gene Name
selenoprotein P
Gene Aliases
SEPP1
Chromosome Location
Chr.21: 24465317 - 24475428 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135605.1 NP_001129077.1 4 - 5 521 103

View Full Product Details