Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PMEL
Entrez Gene ID
100033885
Gene Name
premelanosome protein
Gene Aliases
PMEL17, SILV
Chromosome Location
Chr.6: 74569524 - 74578149 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163889.1 NP_001157361.1 9 - 10 1763 113
XM_005611061.3 XP_005611118.1 - 1990 113
XM_023642149.1 XP_023497917.1 - 2223 113
XM_023642150.1 XP_023497918.1 - 2070 113
GenBank mRNA AF076780.1 - 5 - 6 1183 113

View Full Product Details