Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAPPA
Entrez Gene ID
100033920
Gene Name
pappalysin 1
Gene Aliases
PAPP-A
Chromosome Location
Chr.25: 21035849 - 21279423 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_023628753.1 XP_023484521.1 - 2345 91
GenBank mRNA AF421143.1 - 1 - 2 160 91

View Full Product Details