Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHIT1
Entrez Gene ID
100065255
Gene Name
chitinase 1
Gene Aliases
-
Chromosome Location
Chr.30: 31307357 - 31319870 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143797.1 NP_001137269.1 9 - 10 1105 71
GenBank mRNA EF694759.1 - 9 - 10 1105 71

View Full Product Details