Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACP5
Entrez Gene ID
100056621
Gene Name
acid phosphatase 5, tartrate resistant
Gene Aliases
-
Chromosome Location
Chr.7: 50593391 - 50597109 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246672.1 NP_001233601.1 3 - 4 449 69
XM_005611907.3 XP_005611964.1 - 811 69
XM_005611908.3 XP_005611965.1 - 602 69
XM_023644136.1 XP_023499904.1 - 603 69
GenBank mRNA JN638567.1 - 3 - 4 449 69

View Full Product Details