Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SELENOK
Entrez Gene ID
100316857
Gene Name
selenoprotein K
Gene Aliases
SELK
Chromosome Location
Chr.16: 35504282 - 35511190 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168396.1 NP_001161868.1 4 - 5 451 70
GenBank mRNA CX598013.1 - 4 - 5 332 70

View Full Product Details