Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPATCH11
Entrez Gene ID
421468
Gene Name
G-patch domain containing 11
Gene Aliases
CCDC75
Chromosome Location
Chr.3: 32120011 - 32134373 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277661.1 NP_001264590.1 4 - 5 581 149
NM_001277662.1 NP_001264591.1 4 - 5 581 149
XM_025148695.1 XP_025004463.1 4 - 5 584 149
XM_025148696.1 XP_025004464.1 4 - 5 585 149
XM_025148697.1 XP_025004465.1 4 - 5 585 149
XM_025148698.1 XP_025004466.1 4 - 5 585 149
GenBank mRNA BU111783.1 - 3 - 4 388 149
BU225506.1 - 4 - 5 563 149
BU371798.1 - 3 - 4 175 149
BU441924.1 - 2 - 3 149 150
BX929417.1 - 4 - 5 582 149
BX932819.2 - 4 - 5 563 149

View Full Product Details