Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP26
Entrez Gene ID
426921
Gene Name
dual specificity phosphatase 26 (putative)
Gene Aliases
-
Chromosome Location
Chr.22: 1926344 - 1930743 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015297432.2 XP_015152918.1 2 - 3 183 73
XM_025142886.1 XP_024998654.1 2 - 3 190 73
XM_424530.5 XP_424530.1 1 - 2 277 73
GenBank mRNA BX929514.1 - 1 - 2 58 73
BX931136.2 - 1 - 2 33 73

View Full Product Details