Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERHL
Entrez Gene ID
417976
Gene Name
serine hydrolase-like (pseudogene)
Gene Aliases
SERHL2
Chromosome Location
Chr.1: 49205225 - 49213440 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277373.1 NP_001264302.1 7 - 8 655 64
XM_015285070.2 XP_015140556.1 6 - 7 544 64
XM_025148464.1 XP_025004232.1 7 - 8 766 64
GenBank mRNA BX929989.1 - 6 - 7 562 64
BX932388.1 - 6 - 7 542 64
BX935138.2 - - 555 64
CD218551.1 - 7 - 8 624 64
DN929581.1 - 5 - 6 281 64

View Full Product Details