Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERHL
Entrez Gene ID
417976
Gene Name
serine hydrolase-like (pseudogene)
Gene Aliases
SERHL2
Chromosome Location
Chr.1: 49205225 - 49213440 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX929989.1 - 7 - 8 648 101

View Full Product Details